Assay Design



Gene Transcripts

Gene Symbol
Naaladl2
Entrez Gene ID
100761363
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613891: 39938 - 901079 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642941.1 XP_007641131.1 14 - 15 2064 107

View Full Product Details