Assay Design



Gene Transcripts

Gene Symbol
Fam83h
Entrez Gene ID
100754245
Gene Name
family with sequence similarity 83, member H
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614411: 754072 - 766897 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507632.2 XP_003507680.1 4 - 5 813 66

View Full Product Details