Assay Design



Gene Transcripts

Gene Symbol
LOC100751590
Entrez Gene ID
100751590
Gene Name
chromosome unknown open reading frame, human C11orf83
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614577: 191282 - 191919 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508733.2 XP_003508781.1 1 - 2 214 86

View Full Product Details