Assay Design



Gene Transcripts

Gene Symbol
Dhh
Entrez Gene ID
100768865
Gene Name
desert hedgehog
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614949: 99735 - 104266 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510746.1 XP_003510794.1 2 - 3 564 91

View Full Product Details