Assay Design



Gene Transcripts

Gene Symbol
Tprn
Entrez Gene ID
100750447
Gene Name
taperin
Gene Aliases
I79_022716
Chromosome Location
Chr.Un|NW_003616080: 68737 - 76575 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653948.1 XP_007652138.1 2 - 3 1334 91
GenBank mRNA JI869730.1 - 1 - 2 108 91

View Full Product Details