Assay Design



Gene Transcripts

Gene Symbol
Rxra
Entrez Gene ID
100768377
Gene Name
retinoid X receptor, alpha
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615693: 103968 - 130918 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653390.1 XP_007651580.1 8 - 9 1233 97
GenBank mRNA JI879460.1 - 4 - 5 451 97
JP049010.1 - 4 - 5 451 97

View Full Product Details