Assay Design



Gene Transcripts

Gene Symbol
Tmem89
Entrez Gene ID
100760698
Gene Name
transmembrane protein 89
Gene Aliases
I79_011271
Chromosome Location
Chr.Un|NW_003614047: 114971 - 115848 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504236.1 XP_003504284.1 1 - 2 292 63

View Full Product Details