Assay Design



Gene Transcripts

Gene Symbol
Kcnc2
Entrez Gene ID
100759610
Gene Name
potassium voltage-gated channel, Shaw-related subfamily, member 2
Gene Aliases
I79_008985
Chromosome Location
Chr.Un|NW_003613895: 890270 - 1076038 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502413.1 XP_003502461.1 2 - 3 1616 83
XM_003502414.1 XP_003502462.1 2 - 3 1616 83

View Full Product Details