Assay Design



Gene Transcripts

Gene Symbol
Manbal
Entrez Gene ID
100756897
Gene Name
mannosidase, beta A, lysosomal-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613641: 1185307 - 1214342 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497097.2 XP_003497145.1 2 - 3 493 73
XM_007607813.1 XP_007606003.1 2 - 3 282 73
XM_007607818.1 XP_007606008.1 2 - 3 290 73
XM_007607824.1 XP_007606014.1 2 - 3 270 73
GenBank mRNA JI896035.1 - 2 - 3 522 73
JP055842.1 - 2 - 3 522 73

View Full Product Details