Assay Design



Gene Transcripts

Gene Symbol
Armc5
Entrez Gene ID
100772037
Gene Name
armadillo repeat containing 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616970: 79530 - 86734 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514790.2 XP_003514838.1 3 - 4 1435 74

View Full Product Details