Assay Design



Gene Transcripts

Gene Symbol
Ccni
Entrez Gene ID
100752774
Gene Name
cyclin I
Gene Aliases
Cyclin-I
Chromosome Location
Chr.Un|NW_003615482: 115990 - 145973 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652989.1 XP_007651179.1 3 - 4 318 107
GenBank mRNA JI862911.1 - 3 - 4 321 107
JP060511.1 - 3 - 4 321 107

View Full Product Details