Assay Design



Gene Transcripts

Gene Symbol
Pef1
Entrez Gene ID
100759974
Gene Name
penta-EF-hand domain containing 1
Gene Aliases
Peflin
Chromosome Location
Chr.Un|NW_003613689: 205064 - 224130 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007626479.1 XP_007624669.1 3 - 4 470 73
GenBank mRNA JI878707.1 - 2 - 3 260 73

View Full Product Details