Assay Design



Gene Transcripts

Gene Symbol
Calm3
Entrez Gene ID
100762749
Gene Name
calmodulin 3 (phosphorylase kinase, delta)
Gene Aliases
Calmodulin, I79_018847
Chromosome Location
Chr.Un|NW_003614899: 181914 - 184507 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651153.1 XP_007649343.1 1 - 2 73 66
GenBank mRNA JI893250.1 - 1 - 2 73 66

View Full Product Details