Assay Design



Gene Transcripts

Gene Symbol
Ccdc97
Entrez Gene ID
100773067
Gene Name
coiled-coil domain containing 97
Gene Aliases
I79_020427
Chromosome Location
Chr.Un|NW_003615239: 145796 - 151951 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003511815.2 XP_003511863.1 4 - 5 882 70

View Full Product Details