Assay Design



Gene Transcripts

Gene Symbol
Mstn
Entrez Gene ID
100770972
Gene Name
myostatin
Gene Aliases
I79_023865
Chromosome Location
Chr.Un|NW_003616762: 83346 - 88095 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514604.2 XP_003514652.1 1 - 2 373 70

View Full Product Details