Assay Design



Gene Transcripts

Gene Symbol
Oscar
Entrez Gene ID
100764692
Gene Name
osteoclast associated, immunoglobulin-like receptor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614747: 406293 - 412597 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509653.1 XP_003509701.1 1 - 2 41 68

View Full Product Details