Assay Design



Gene Transcripts

Gene Symbol
Gjb4
Entrez Gene ID
100765131
Gene Name
gap junction protein, beta 4, 30.3kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613817: 441509 - 451407 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641588.1 XP_007639778.1 1 - 2 1066 83

View Full Product Details