Assay Design



Gene Transcripts

Gene Symbol
Nck2
Entrez Gene ID
100762895
Gene Name
NCK adaptor protein 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613742: 1412157 - 1533195 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640280.1 XP_007638470.1 3 - 4 509 57
XM_007640281.1 XP_007638471.1 3 - 4 466 57
XM_007640282.1 XP_007638472.1 3 - 4 466 57
XM_007640283.1 XP_007638473.1 3 - 4 506 57

View Full Product Details