Assay Design



Gene Transcripts

Gene Symbol
Fam53a
Entrez Gene ID
100766117
Gene Name
family with sequence similarity 53, member A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614033: 423170 - 455276 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644626.1 XP_007642816.1 2 - 3 819 71
XM_007644627.1 XP_007642817.1 2 - 3 823 71
XM_007644628.1 XP_007642818.1 3 - 4 925 71
XM_007644629.1 XP_007642819.1 2 - 3 545 71
XM_007644630.1 XP_007642820.1 2 - 3 286 71
XM_007644631.1 XP_007642821.1 2 - 3 819 71

View Full Product Details