Assay Design



Gene Transcripts

Gene Symbol
Trim68
Entrez Gene ID
100760957
Gene Name
tripartite motif containing 68
Gene Aliases
I79_024164
Chromosome Location
Chr.Un|NW_003617006: 36547 - 48192 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514821.2 XP_003514869.1 6 - 7 1052 98
XM_007654875.1 XP_007653065.1 6 - 7 1015 98

View Full Product Details