Assay Design



Gene Transcripts

Gene Symbol
Slc22a16
Entrez Gene ID
100753777
Gene Name
solute carrier family 22 (organic cation/carnitine transporter), member 16
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613639: 3278947 - 3315805 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497006.1 XP_003497054.1 3 - 4 657 91

View Full Product Details