Assay Design



Gene Transcripts

Gene Symbol
Tmem52
Entrez Gene ID
100755990
Gene Name
transmembrane protein 52
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614679: 124609 - 125955 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509281.1 XP_003509329.1 1 - 2 69 83
XM_007649855.1 XP_007648045.1 1 - 2 69 83
XM_007649856.1 XP_007648046.1 1 - 2 69 83

View Full Product Details