Assay Design



Gene Transcripts

Gene Symbol
Hbe1
Entrez Gene ID
100768374
Gene Name
hemoglobin, epsilon 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615521: 28442 - 29716 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512618.2 XP_003512666.1 2 - 3 318 76

View Full Product Details