Assay Design



Gene Transcripts

Gene Symbol
Efna2
Entrez Gene ID
100755670
Gene Name
ephrin-A2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613904: 962124 - 971853 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502612.2 XP_003502660.1 3 - 4 400 84

View Full Product Details