Assay Design



Gene Transcripts

Gene Symbol
Hebp1
Entrez Gene ID
100752514
Gene Name
heme binding protein 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613840: 1747814 - 1773293 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501269.2 XP_003501317.1 1 - 2 179 67
GenBank mRNA JI874658.1 - 1 - 2 171 67
JP044806.1 - 1 - 2 172 67

View Full Product Details