Assay Design



Gene Transcripts

Gene Symbol
Tppp2
Entrez Gene ID
100758674
Gene Name
tubulin polymerization-promoting protein family member 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614416: 774623 - 776541 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507718.2 XP_003507766.2 1 - 2 175 73

View Full Product Details