Assay Design



Gene Transcripts

Gene Symbol
Gphb5
Entrez Gene ID
100762212
Gene Name
glycoprotein hormone beta 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613728: 1228299 - 1231029 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499303.1 XP_003499351.1 1 - 2 199 78

View Full Product Details