Assay Design
Gene Transcripts
- Gene Symbol
- Serpina11
- Entrez Gene ID
-
100765747
- Gene Name
- serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11
- Gene Aliases
-
I79_017526
- Chromosome Location
-
Chr.Un|NW_003614700: 482213 - 490188 on Build CriGri-PICRH-1.0
View Full Product Details