Assay Design



Gene Transcripts

Gene Symbol
Serpina11
Entrez Gene ID
100765747
Gene Name
serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 11
Gene Aliases
I79_017526
Chromosome Location
Chr.Un|NW_003614700: 482213 - 490188 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509381.1 XP_003509429.1 2 - 3 919 91

View Full Product Details