Assay Design



Gene Transcripts

Gene Symbol
Rilpl2
Entrez Gene ID
100774122
Gene Name
Rab interacting lysosomal protein-like 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614852: 39118 - 48364 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510262.2 XP_003510310.1 3 - 4 834 81
GenBank mRNA JI868163.1 - 2 - 3 199 81
JP064160.1 - 2 - 3 199 81

View Full Product Details