Assay Design



Gene Transcripts

Gene Symbol
Isg20
Entrez Gene ID
100765291
Gene Name
interferon stimulated exonuclease gene 20kDa
Gene Aliases
I79_021329
Chromosome Location
Chr.Un|NW_003615490: 226448 - 240497 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512537.2 XP_003512585.1 2 - 3 429 73

View Full Product Details