Assay Design



Gene Transcripts

Gene Symbol
Fabp9
Entrez Gene ID
100761969
Gene Name
fatty acid binding protein 9, testis
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614824: 249677 - 252805 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510078.1 XP_003510126.1 2 - 3 242 103

View Full Product Details