Assay Design



Gene Transcripts

Gene Symbol
Zfp14
Entrez Gene ID
100759956
Gene Name
ZFP14 zinc finger protein
Gene Aliases
I79_023277
Chromosome Location
Chr.Un|NW_003616361: 86152 - 91931 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654323.1 XP_007652513.1 3 - 3 2019 124
GenBank mRNA JP059190.1 - 1 - 1 383 124

View Full Product Details