Assay Design



Gene Transcripts

Gene Symbol
Hs6st3
Entrez Gene ID
100765877
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Gene Aliases
I79_000390
Chromosome Location
Chr.Un|NW_003613586: 1586999 - 1593007 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495239.1 XP_003495287.1 2 - 2 622 94

View Full Product Details