Assay Design



Gene Transcripts

Gene Symbol
Mblac1
Entrez Gene ID
100770291
Gene Name
metallo-beta-lactamase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614875: 211843 - 212736 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651047.1 XP_007649237.1 1 - 1 375 105

View Full Product Details