Assay Design



Gene Transcripts

Gene Symbol
Dnajc30
Entrez Gene ID
103162154
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 30
Gene Aliases
I79_007245
Chromosome Location
Chr.Un|NW_003613822: 38792 - 40593 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641678.1 XP_007639868.1 2 - 2 221 106

View Full Product Details