Assay Design



Gene Transcripts

Gene Symbol
Cpxcr1
Entrez Gene ID
100768537
Gene Name
CPX chromosome region, candidate 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614156: 46317 - 47282 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505506.1 XP_003505554.1 2 - 2 716 111

View Full Product Details