Assay Design



Gene Transcripts

Gene Symbol
Tpt1
Entrez Gene ID
100770904
Gene Name
tumor protein, translationally-controlled 1
Gene Aliases
I79_006528
Chromosome Location
Chr.Un|NW_003613785: 171650 - 173756 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641010.1 XP_007639200.1 1 - 1 726 106

View Full Product Details