Assay Design



Gene Transcripts

Gene Symbol
Blcap
Entrez Gene ID
100757663
Gene Name
bladder cancer associated protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613641: 1380148 - 1390482 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497100.2 XP_003497148.1 2 - 2 2026 93
XM_007607858.1 XP_007606048.1 2 - 2 1958 93

View Full Product Details