Assay Design



Gene Transcripts

Gene Symbol
LIF
Entrez Gene ID
102188524
Gene Name
LIF interleukin 6 family cytokine
Gene Aliases
-
Chromosome Location
Chr.17: 1999634 - 2005932 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691625.3 XP_005691682.2 2 - 3 354 76

View Full Product Details