Assay Design



Gene Transcripts

Gene Symbol
TACR2
Entrez Gene ID
102190490
Gene Name
tachykinin receptor 2
Gene Aliases
-
Chromosome Location
Chr.28: 19890748 - 19902387 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699135.3 XP_005699192.1 3 - 4 1202 77

View Full Product Details