Assay Design



Gene Transcripts

Gene Symbol
OAZ1
Entrez Gene ID
100860946
Gene Name
ornithine decarboxylase antizyme 1
Gene Aliases
-
Chromosome Location
Chr.7: 89278871 - 89282375 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285559.1 NP_001272488.1 2 - 3 445 125
GenBank mRNA GU396146.1 - 2 - 3 350 125
HQ326240.1 - 2 - 3 349 125
JO579793.1 - 2 - 3 445 125
JO634402.2 - 2 - 3 379 125
KA318171.1 - 3 - 4 822 125

View Full Product Details