Assay Design



Gene Transcripts

Gene Symbol
AAR2
Entrez Gene ID
102177640
Gene Name
AAR2 splicing factor
Gene Aliases
-
Chromosome Location
Chr.13: 64931142 - 64951036 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688516.3 XP_005688573.2 3 - 4 1099 79
XM_005688517.3 XP_005688574.2 3 - 4 1137 79
XM_018057806.1 XP_017913295.1 2 - 3 1026 79
GenBank mRNA JO438592.2 - 1 - 2 111 79

View Full Product Details