Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005692435.3 | XP_005692492.1 | 3 - 4 | 451 | 58 |
XM_013971329.2 | XP_013826783.1 | 3 - 4 | 461 | 58 | |
GenBank mRNA | JO441518.2 | - | 1 - 2 | 589 | 58 |
JO616024.2 | - | 3 - 4 | 433 | 58 |