Assay Design



Gene Transcripts

Gene Symbol
TIMP1
Entrez Gene ID
100860924
Gene Name
TIMP metallopeptidase inhibitor 1
Gene Aliases
TIMP-1
Chromosome Location
Chr.X|NW_017189516: 20056637 - 20060291 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314350.1 NP_001301279.1 4 - 5 477 75
XM_005700836.3 XP_005700893.2 5 - 6 671 75
GenBank mRNA GU446671.1 - 4 - 5 386 75
JO647226.2 - 1 - 2 114 75
KM215210.1 - 4 - 5 477 75

View Full Product Details