Assay Design



Gene Transcripts

Gene Symbol
KCNK17
Entrez Gene ID
102185792
Gene Name
potassium two pore domain channel subfamily K member 17
Gene Aliases
-
Chromosome Location
Chr.23: 35645363 - 35659153 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039322.1 XP_017894811.1 2 - 3 492 69

View Full Product Details