Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_013966938.2 | XP_013822392.2 | 2 - 3 | 248 | 71 |
XM_018054526.1 | XP_017910015.1 | 3 - 4 | 630 | 71 | |
XM_018054527.1 | XP_017910016.1 | 3 - 4 | 416 | 71 | |
XM_018054528.1 | XP_017910017.1 | 3 - 4 | 567 | 71 | |
XM_018054529.1 | XP_017910018.1 | 3 - 4 | 551 | 71 | |
XM_018054530.1 | XP_017910019.1 | 2 - 3 | 482 | 71 | |
GenBank mRNA | JO413718.2 | - | 2 - 3 | 440 | 71 |