Assay Design



Gene Transcripts

Gene Symbol
POLE3
Entrez Gene ID
102173090
Gene Name
DNA polymerase epsilon 3, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.8: 102363417 - 102366295 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684327.3 XP_005684384.1 1 - 2 364 110
GenBank mRNA JP351500.2 - 1 - 2 390 110

View Full Product Details