Assay Design



Gene Transcripts

Gene Symbol
TSEN15
Entrez Gene ID
102173592
Gene Name
tRNA splicing endonuclease subunit 15
Gene Aliases
-
Chromosome Location
Chr.16: 63723675 - 63754233 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690969.3 XP_005691026.3 2 - 3 290 86
GenBank mRNA JO603770.2 - 2 - 3 294 86

View Full Product Details