Assay Design



Gene Transcripts

Gene Symbol
FBXO39
Entrez Gene ID
102190351
Gene Name
F-box protein 39
Gene Aliases
-
Chromosome Location
Chr.19: 24946788 - 24954195 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064067.1 XP_017919556.1 2 - 3 1150 64

View Full Product Details